Assay Details
Target Gene Details
Entrez Gene ID: | 8883 |
Gene Name: | NEDD8 activating enzyme E1 subunit 1 |
Gene Aliases: |
A-116A10.1, APPBP1, HPP1, ula-1 |
Location: |
Chr.16:66802878-66830977 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAE1 | NM_001018159.1 | 21 | 1828 | NP_001018169.1 |
NM_001018160.1 | 19 | 1626 | NP_001018170.1 | |
NM_001286500.1 | 20 | 1739 | NP_001273429.1 | |
NM_003905.3 | 20 | 1730 | NP_003896.1 | |
XM_005256215.1 | 19 | 1700 | XP_005256272.1 | |
XM_011523422.2 | 18 | 1542 | XP_011521724.1 | |
AK097680.1 | 20 | 1739 | ||
AK298159.1 | 21 | 1777 | ||
AK312784.1 | ||||
AL136798.1 | 20 | 1699 | CAB66732.1 | |
AY197612.1 | 20 | 1668 | AAP35030.1 | |
BC000480.2 | 20 | 1661 | AAH00480.1 | |
BC013301.2 | 20 | 1661 | AAH13301.1 | |
U50939.1 | 20 | 1726 | AAC50477.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |