Assay Details
Target Gene Details
Entrez Gene ID: | 3265 |
Gene Name: | HRas proto-oncogene, GTPase |
Gene Aliases: |
C-BAS/HAS, C-H-RAS, C-HA-RAS1, CTLO, H-RASIDX, HAMSV, HRAS1, RASH1, p21ras |
Location: |
Chr.11:532242-535567 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HRAS | NM_001130442.2 | 5 | 769 | NP_001123914.1 |
NM_001318054.1 | NP_001304983.1 | |||
NM_005343.3 | NP_005334.1 | |||
NM_176795.4 | 6 | 851 | NP_789765.1 | |
AB451336.1 | ||||
AB451485.1 | ||||
AF493916.1 | AAM12630.1 | |||
BC006499.2 | AAH06499.1 | |||
BC095471.1 | AAH95471.1 | |||
BM801600.1 | ||||
BM801632.1 | ||||
BQ574535.1 | 1 | 418 | ||
BQ674260.1 | 5 | 709 | ||
BT019421.1 | AAV38228.1 | |||
CR536579.1 | CAG38816.1 | |||
CR542271.1 | CAG47067.1 | |||
KJ534867.1 | ||||
KJ534868.1 |
Target Gene Details
Entrez Gene ID: | 115399 |
Gene Name: | leucine rich repeat containing 56 |
Gene Aliases: |
- |
Location: |
Chr.11:518967-554916 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LRRC56 | XM_011519875.2 | XP_011518177.1 | ||
XM_011519877.2 | XP_011518179.1 | |||
XM_017017167.1 | XP_016872656.1 | |||
XM_017017168.1 | XP_016872657.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv467634 | Chr.11:511805 - 619789 on Build GRCh38 | Loss | MIR210 LRRC56 RASSF7 HRAS LOC101059906 MIR210HG LMNTD2 CDHR5 IRF7 LOC143666 PHRF1 |
nsv527327 | Chr.11:295343 - 620599 on Build GRCh38 | Loss | RASSF7 RNH1 CDHR5 SIGIRR IFITM1 PGGHG MIR210 LRRC56 IFITM5 B4GALNT4 HRAS LOC101059906 IFITM3 MIR210HG LMNTD2 PKP3 PTDSS2 ANO9 IFITM2 IRF7 LOC143666 PHRF1 |
nsv951270 | Chr.11:528401 - 628100 on Build GRCh38 | Deletion | MIR210 LRRC56 RASSF7 HRAS LOC101059906 MIR210HG LMNTD2 CDHR5 IRF7 SCT LOC143666 PHRF1 |
nsv469923 | Chr.11:378188 - 1302334 on Build GRCh38 | Loss | DRD4 MUC6 RPLP2 EPS8L2 SIGIRR CD151 LRRC56 CRACR2B HRAS LOC101059906 DEAF1 MIR210HG SLC25A22 LOC101927503 PTDSS2 ANO9 IRF7 MUC5AC LOC143666 PHRF1 TOLLIP RASSF7 CEND1 RNH1 PIDD1 CDHR5 SNORA52 TSPAN4 MUC2 LOC171391 PANO1 MIR210 MUC5B TMEM80 B4GALNT4 LMNTD2 PNPLA2 PKP3 AP2A2 POLR2L CHID1 PDDC1 TALDO1 SCT MIR6744 |
esv2759794 | Chr.11:409846 - 1336162 on Build GRCh38 | Loss | DRD4 MUC6 RPLP2 EPS8L2 SIGIRR CD151 LRRC56 CRACR2B HRAS LOC101059906 DEAF1 MIR210HG SLC25A22 LOC101927503 PTDSS2 ANO9 IRF7 MUC5AC LOC143666 PHRF1 TOLLIP-AS1 TOLLIP RASSF7 CEND1 RNH1 PIDD1 CDHR5 SNORA52 TSPAN4 MUC2 LOC171391 PANO1 MIR210 MUC5B TMEM80 LMNTD2 PNPLA2 AP2A2 POLR2L CHID1 PDDC1 TALDO1 SCT MIR6744 |
nsv1053964 | Chr.11:491334 - 692433 on Build GRCh38 | Gain | DRD4 RASSF7 RNH1 CDHR5 MIR210 LRRC56 HRAS LOC101059906 DEAF1 MIR210HG LMNTD2 PTDSS2 IRF7 SCT LOC143666 PHRF1 |
dgv182e199 | Chr.11:349830 - 943372 on Build GRCh38 | Deletion | DRD4 RPLP2 EPS8L2 SIGIRR CD151 LRRC56 CRACR2B HRAS LOC101059906 DEAF1 MIR210HG SLC25A22 PTDSS2 ANO9 IRF7 LOC143666 PHRF1 RASSF7 CEND1 RNH1 PIDD1 CDHR5 SNORA52 TSPAN4 LOC171391 PANO1 MIR210 TMEM80 B4GALNT4 LMNTD2 PNPLA2 PKP3 AP2A2 POLR2L CHID1 PDDC1 TALDO1 SCT |
nsv1159791 | Chr.11:461793 - 909492 on Build GRCh38 | Deletion | DRD4 RPLP2 EPS8L2 CD151 LRRC56 CRACR2B HRAS LOC101059906 DEAF1 MIR210HG SLC25A22 PTDSS2 IRF7 LOC143666 PHRF1 RASSF7 CEND1 RNH1 PIDD1 CDHR5 SNORA52 TSPAN4 LOC171391 PANO1 MIR210 TMEM80 LMNTD2 PNPLA2 POLR2L CHID1 PDDC1 TALDO1 SCT |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |