Assay Details
Target Gene Details
Entrez Gene ID: | 4728 |
Gene Name: | NADH:ubiquinone oxidoreductase core subunit S8 |
Gene Aliases: |
CI-23k, CI23KD, TYKY |
Location: |
Chr.11:68030617-68036647 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFS8 | NM_002496.3 | NP_002487.1 | ||
XM_005274013.1 | XP_005274070.1 | |||
XM_005274014.2 | XP_005274071.1 | |||
XM_011545053.2 | XP_011543355.1 | |||
AK223114.1 | BAD96834.1 | |||
AK314546.1 | ||||
BC119753.1 | ||||
BC119754.2 | ||||
DB498707.1 | ||||
U65579.1 | AAB51776.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv8836 | Chr.11:67985408 - 68052332 on Build GRCh38 | Gain | TCIRG1 NDUFS8 MIR6753 MIR4691 UNC93B1 ALDH3B1 MIR7113 |
dgv1981n54 | Chr.11:67579272 - 68049469 on Build GRCh38 | Gain | ALDH3B2 TCIRG1 NDUFS8 MIR4691 UNC93B1 TBX10 GSTP1 ALDH3B1 MIR7113 NUDT8 LOC105369360 MIR6753 ACY3 C11orf72 FAM86C2P DOC2GP NDUFV1 |
nsv825963 | Chr.11:67996205 - 68429586 on Build GRCh38 | Gain | TCIRG1 NDUFS8 MIR6753 C11orf24 LRP5 CHKA MIR4691 UNC93B1 KMT5B ALDH3B1 MIR7113 |
nsv555282 | Chr.11:68031100 - 68050858 on Build GRCh38 | Loss | TCIRG1 NDUFS8 MIR6753 MIR4691 MIR7113 |
dgv1983n54 | Chr.11:68022085 - 68053569 on Build GRCh38 | Loss | TCIRG1 NDUFS8 MIR6753 CHKA MIR4691 ALDH3B1 MIR7113 |
esv2759835 | Chr.11:67705434 - 68123822 on Build GRCh38 | Gain+Loss | TCIRG1 NDUFS8 LOC105369360 MIR6753 CHKA MIR4691 UNC93B1 FAM86C2P ALDH3B1 MIR7113 |
nsv951026 | Chr.11:67994030 - 68053133 on Build GRCh38 | Deletion | TCIRG1 NDUFS8 MIR6753 CHKA MIR4691 UNC93B1 ALDH3B1 MIR7113 |
nsv469964 | Chr.11:67997585 - 68053569 on Build GRCh38 | Loss | TCIRG1 NDUFS8 MIR6753 CHKA MIR4691 UNC93B1 ALDH3B1 MIR7113 |
nsv468606 | Chr.11:67585218 - 68049469 on Build GRCh38 | Gain | ALDH3B2 TCIRG1 NDUFS8 MIR4691 UNC93B1 TBX10 GSTP1 ALDH3B1 MIR7113 NUDT8 LOC105369360 MIR6753 ACY3 C11orf72 FAM86C2P DOC2GP NDUFV1 |
dgv1221n100 | Chr.11:67979689 - 68054143 on Build GRCh38 | Gain | TCIRG1 NDUFS8 MIR6753 CHKA MIR4691 UNC93B1 ALDH3B1 MIR7113 |
nsv469856 | Chr.11:67871566 - 68092023 on Build GRCh38 | Gain | TCIRG1 NDUFS8 LOC105369360 MIR6753 CHKA MIR4691 UNC93B1 ALDH3B1 MIR7113 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |