Assay Details
Target Gene Details
Entrez Gene ID: | 19989 |
Gene Name: | ribosomal protein L7 |
Gene Aliases: |
Rpl7a, Surf-3 |
Location: |
Chr.1:16101295-16104433 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rpl7 | NM_011291.5 | NP_035421.2 | ||
AK017074.1 | ||||
AK050656.1 | BAC34366.1 | |||
AK088291.1 | BAC40262.1 | |||
AK135460.1 | ||||
AK146219.1 | BAE26987.1 | |||
AK150943.1 | BAE29978.1 | |||
AK167124.1 | BAE39272.1 | |||
AK168137.1 | BAE40105.1 | |||
AK168227.1 | ||||
BB839528.1 | ||||
BC025909.1 | AAH25909.1 | |||
BC051261.1 | AAH51261.1 | |||
BC096452.1 | AAH96452.1 | |||
M29016.1 | AAA40070.1 | |||
M85235.1 | AAA40064.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic Validated |