Assay Details
Target Gene Details
Entrez Gene ID: | 12799 |
Gene Name: | 2',3'-cyclic nucleotide 3' phosphodiesterase |
Gene Aliases: |
CNPase, Cnp-1, Cnp1 |
Location: |
Chr.11:100574909-100591521 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cnp | NM_001146318.1 | 1 | 132 | NP_001139790.1 |
NM_009923.2 | NP_034053.2 | |||
AK041969.1 | 1 | 133 | ||
AK050628.1 | BAC34351.1 | |||
AK087415.1 | ||||
AK140280.1 | 1 | 133 | BAE24314.1 | |
AK154654.1 | BAE32745.1 | |||
AK158318.1 | BAE34457.1 | |||
AK158520.1 | BAE34543.1 | |||
AK172318.1 | ||||
AK172514.1 | ||||
BC005544.1 | AAH05544.1 | |||
BC021904.1 | AAH21904.1 | |||
BY333311.1 | ||||
M31810.1 | 1 | 499 | AAA37429.1 | |
M58045.1 | AAA37430.1 |
More Information
Set Membership: |
Intragenic Exonic Validated |