Assay Details
Target Gene Details
Entrez Gene ID: | 246277 |
Gene Name: | cysteine sulfinic acid decarboxylase |
Gene Aliases: |
Csd |
Location: |
Chr.15:102176997-102204231 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Csad | NM_144942.4 | 15 | 1676 | NP_659191.1 |
AK005015.1 | 15 | 1677 | BAB23747.1 | |
AY033912.1 | 15 | 1593 | AAK60398.1 |
More Information
Set Membership: |
Intragenic Exonic |