Assay Details
Target Gene Details
Entrez Gene ID: | 56794 |
Gene Name: | 2-hydroxyacyl-CoA lyase 1 |
Gene Aliases: |
1600020H07Rik, Hpcl, Phyh2 |
Location: |
Chr.14:31607226-31641286 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hacl1 | NM_019975.3 | 17 | 2229 | NP_064359.2 |
AK041686.1 | 17 | 2225 | BAC31032.1 |
More Information
Set Membership: |
Intragenic Exonic |