Assay Details
Target Gene Details
Entrez Gene ID: | 70192 |
Gene Name: | CD209g antigen |
Gene Aliases: |
2310066I10Rik, AI481039, SIGNR7 |
Location: |
Chr.8:4134246-4137707 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cd209g | NM_027343.3 | 8 | 723 | NP_081619.3 |
AI606925.1 | ||||
AK010071.1 | 7 | 684 | ||
AM286437.1 | 8 | 726 | ||
BC118542.1 | 1 | 3248 | AAI18543.1 |
More Information
Set Membership: |
Intragenic Exonic |