Assay Details
Target Gene Details
Entrez Gene ID: | 235459 |
Gene Name: | general transcription factor II A, 2 |
Gene Aliases: |
TFIIA-gamma, TfIIg, Tfiia2 |
Location: |
Chr.9:70011851-70022866 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gtf2a2 | NM_001039519.2 | 5 | 826 | NP_001034608.1 |
NM_001282083.1 | 5 | 1117 | NP_001269012.1 | |
NM_001282084.1 | 4 | 699 | NP_001269013.1 | |
NR_104093.1 | 6 | 965 | ||
AK165466.1 | 5 | 824 | BAE38203.1 | |
CR514895.1 | 1 | 59 |
More Information
Set Membership: |
Intragenic Exonic |