Assay Details
Target Gene Details
Entrez Gene ID: | 353211 |
Gene Name: | prune homolog 2 (Drosophila) |
Gene Aliases: |
6330414G02Rik, A230083H22Rik, A330102H22Rik, Bmcc1, mKIAA0367 |
Location: |
Chr.19:16955670-17223958 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prune2 | NM_181348.4 | 11 | 8563 | NP_851993.3 |
AK038997.1 | 3 | 327 | BAC30198.1 | |
AK054190.1 | 3 | 326 | BAC35688.1 | |
AK163576.1 | 3 | 316 | BAE37405.1 | |
AK165582.1 | 3 | 379 | BAE38273.1 | |
AK170001.1 | 3 | 257 | BAE41506.1 | |
AK172939.1 | 3 | 316 | BAD32217.1 | |
BC094224.1 | 4 | 2807 | AAH94224.1 | |
BC095978.1 | 3 | 301 | AAH95978.1 | |
FR693373.1 | 3 | 325 | ||
GQ403051.1 | 3 | 318 |