Assay Details
Target Gene Details
Entrez Gene ID: | 320865 |
Gene Name: | cadherin 18 |
Gene Aliases: |
B230220E17Rik, Cdh14l |
Location: |
Chr.15:22548899-23474432 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdh18 | AK045672.1 | BAC32451.1 |
More Information
Set Membership: |
Intragenic Non-exonic |