Assay Details
Target Gene Details
Entrez Gene ID: | 75317 |
Gene Name: | PARP1 binding protein |
Gene Aliases: |
4930547N16Rik, AI503644, PARI |
Location: |
Chr.10:88091398-88146945 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Parpbp | NM_029249.2 | NP_083525.1 | ||
AK030970.1 | BAC27197.1 | |||
AK131981.1 | BAE20920.1 | |||
AK137160.1 | ||||
AK150571.1 | BAE29666.1 | |||
AK151268.1 | BAE30255.1 | |||
AK152208.1 | BAE31036.1 | |||
BC070476.1 |