Assay Details
Target Gene Details
Entrez Gene ID: | 192170 |
Gene Name: | eukaryotic translation initiation factor 4A3 |
Gene Aliases: |
2400003O03Rik, Ddx48, eIF4A-III, mKIAA0111 |
Location: |
Chr.11:119288363-119300043 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif4a3 | NM_138669.1 | 1 | 57 | NP_619610.1 |
AK075920.1 | BAC36054.1 | |||
AK150277.1 | 1 | 73 | BAE29433.1 | |
AK151786.1 | 1 | 39 | ||
AK152824.1 | 1 | 73 | BAE31525.1 | |
AK153359.1 | 1 | 39 | BAE31931.1 | |
AK157855.1 | BAE34233.1 | |||
AK167107.1 | 1 | 95 | BAE39256.1 | |
AK168319.1 | BAE40258.1 | |||
AK169350.1 | BAE41100.1 | |||
BC008132.1 | 1 | 57 | AAH08132.1 | |
BC012862.1 | 1 | 45 | AAH12862.1 |