Assay Details
Target Gene Details
Entrez Gene ID: | 380709 |
Gene Name: | spermatogenesis associated 22 |
Gene Aliases: |
Gm882 |
Location: |
Chr.11:73326537-73346434 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Spata22 | NM_001045531.1 | NP_001038996.1 | ||
BC147586.1 | ||||
BC147607.1 |