Assay Details
Target Gene Details
Entrez Gene ID: | 75458 |
Gene Name: | chemokine-like factor |
Gene Aliases: |
1700001C14Rik, 1810018M11Rik, C32, CKLF1, CKLF3, CKLF4, CKLF5, Cklf2, Cklf6, HSPC224, UCK-1 |
Location: |
Chr.8:104250861-104264936 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cklf | NM_001037841.3 | 3 | 1780 | NP_001032930.1 |
AK032759.1 | 3 | 1710 | ||
AK086063.1 | 3 | 1711 | BAC39602.1 |
More Information
Set Membership: |
Intragenic Exonic |