Assay Details
Target Gene Details
Entrez Gene ID: | 20592 |
Gene Name: | lysine (K)-specific demethylase 5D |
Gene Aliases: |
HY, Jarid1d, Smcy |
Location: |
Chr.Y:897660-943813 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kdm5d | NM_011419.3 | 19 | 3084 | NP_035549.1 |
AB221606.1 | 5 | 737 | ||
AF127244.1 | 19 | 2922 | AAD53048.1 | |
AK140971.1 | 19 | 3186 | BAE24534.1 | |
BC059077.1 | 11 | 1899 | AAH59077.1 | |
CA316252.1 |
More Information
Set Membership: |
Intragenic Exonic |