Assay Details
Target Gene Details
Entrez Gene ID: | 59048 |
Gene Name: | C1GALT1-specific chaperone 1 |
Gene Aliases: |
1500002I11Rik, AB030184, C1GalT2, C81205, Cosmc |
Location: |
Chr.X:38630783-38635143 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C1galt1c1 | NM_021550.3 | 2 | 834 | NP_067525.1 |
AB030184.1 | 2 | 746 | BAA92748.1 | |
AB091728.1 | 1 | 662 | BAD13521.1 | |
AK005118.1 | 2 | 764 | ||
AK008040.1 | 2 | 781 | BAB25426.1 | |
AY159320.1 | 2 | 749 | AAN78130.1 | |
BC029909.1 | 2 | 800 | AAH29909.1 |
More Information
Set Membership: |
Intragenic Exonic |