Assay Details
Target Gene Details
Entrez Gene ID: | 66611 |
Gene Name: | RIB43A domain with coiled-coils 1 |
Gene Aliases: |
2610028I09Rik, W08639 |
Location: |
Chr.X:152004580-152018219 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ribc1 | NM_025660.2 | 5 | 483 | NP_079936.1 |
AK011592.1 | 5 | 483 | BAB27721.1 | |
AK017461.1 | 5 | 375 | BAB30754.3 | |
BC050755.1 | 5 | 471 | AAH50755.1 |
More Information
Set Membership: |
Intragenic Exonic |