Assay Details
Target Gene Details
Entrez Gene ID: | 22668 |
Gene Name: | splicing factor 1 |
Gene Aliases: |
BBP, MZFM, WBP4, Zfp162 |
Location: |
Chr.19:6363690-6378038 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sf1 | NM_001110791.1 | 1 | 70 | NP_001104261.1 |
NM_011750.2 | 1 | 70 | NP_035880.2 | |
NR_122118.1 | 1 | 70 | ||
CJ175598.1 | 1 | 71 |
More Information
Set Membership: |
Intragenic Exonic |