Assay Details
Target Gene Details
Entrez Gene ID: | 16814 |
Gene Name: | ladybird homeobox homolog 1 (Drosophila) |
Gene Aliases: |
Lbx1h |
Location: |
Chr.19:45232684-45237435 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lbx1 | NM_010691.5 | 2 | 775 | NP_034821.2 |
AB108497.1 | 2 | 775 | BAC75634.1 | |
AK144562.1 | 2 | 620 | BAE25939.1 | |
BC119177.1 | 2 | 864 | ||
BC120586.1 | 2 | 864 | AAI20587.1 | |
X90829.1 | 1 | 1920 | CAA62343.1 |
More Information
Set Membership: |
Intragenic Exonic |