Assay Details
Target Gene Details
Entrez Gene ID: | 108101 |
Gene Name: | fermitin family homolog 3 (Drosophila) |
Gene Aliases: |
C79673, Kindlin3 |
Location: |
Chr.19:6998958-7019469 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fermt3 | NM_153795.2 | 15 | 2475 | NP_722490.1 |
AK090332.1 | 15 | 2372 | ||
AK142912.1 | 15 | 2476 | ||
AK149737.1 | 14 | 2898 | ||
AK158357.1 | 15 | 2372 | ||
AK169682.1 | 15 | 2386 | BAE41302.1 | |
BC025119.1 | 15 | 2407 | AAH25119.1 | |
BC032204.1 | 15 | 2451 | AAH32204.1 | |
BC066803.1 | 14 | 2757 | AAH66803.1 |
Target Gene Details
Entrez Gene ID: | 107328 |
Gene Name: | tRNA phosphotransferase 1 |
Gene Aliases: |
AU079016, AW045591, Tpt1, Tpt1h |
Location: |
Chr.19:6996131-6999048 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trpt1 | NM_153597.2 | NP_705825.2 | ||
NR_027351.1 | ||||
AK038830.1 | ||||
BC027575.1 | AAH27575.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |