Assay Details
Target Gene Details
Entrez Gene ID: | 19726 |
Gene Name: | regulatory factor X, 3 (influences HLA class II expression) |
Gene Aliases: |
C230093O12Rik, MRFX3 |
Location: |
Chr.19:27761718-28011698 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rfx3 | NM_001166414.1 | 5 | 468 | NP_001159886.1 |
NM_011265.3 | 4 | 509 | NP_035395.2 | |
AK030008.1 | 6 | 541 | BAC26730.1 | |
AK041120.1 | 5 | 569 | BAC30829.1 | |
AK140928.1 | 4 | 392 | ||
BC017598.1 | 4 | 483 | AAH17598.1 |
More Information
Set Membership: |
Intragenic Exonic |