Assay Details
Target Gene Details
Entrez Gene ID: | 56736 |
Gene Name: | ring finger protein 14 |
Gene Aliases: |
2310075C09Rik, 2610005D23Rik, AA986456, AU041447, D18Ertd188e, D7Bwg0165e, Triad2 |
Location: |
Chr.18:38296635-38317852 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rnf14 | NM_001164621.1 | 6 | 1316 | NP_001158093.1 |
NM_001164622.1 | 4 | 851 | NP_001158094.1 | |
NM_020012.2 | 5 | 1003 | NP_064396.2 | |
AF249667.1 | 5 | 949 | AAF74266.1 | |
AF249668.1 | 3 | 534 | AAF74267.1 | |
AK011316.2 | 6 | 1004 | BAB27541.1 | |
AK038986.1 | 4 | 793 | ||
BC054841.1 | 5 | 933 | AAH54841.1 | |
BC094250.1 | 5 | 954 | AAH94250.1 |
More Information
Set Membership: |
Intragenic Exonic |