Assay Details
Target Gene Details
Entrez Gene ID: | 381218 |
Gene Name: | RIKEN cDNA 4430402I18 gene |
Gene Aliases: |
AW122618, Spata6l |
Location: |
Chr.19:28845528-28970888 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4430402I18Rik | NM_198651.2 | 13 | 1624 | NP_941053.2 |
AK014471.1 | 4 | 402 | ||
AK031997.1 | 13 | 1625 | BAC27642.1 |