Assay Details
Target Gene Details
Entrez Gene ID: | 332396 |
Gene Name: | potassium channel, subfamily K, member 18 |
Gene Aliases: |
Gm781, Tresk, Tresk-2, Trik |
Location: |
Chr.19:59219648-59237370 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kcnk18 | NM_207261.3 | 1 | 281 | NP_997144.1 |
AK139600.1 | 1 | 715 | BAE24080.1 | |
AK162136.1 | 1 | 284 | BAE36746.1 | |
AY325301.1 | 1 | 195 | AAQ91836.1 | |
AY542902.1 | 1 | 195 | AAS48426.1 | |
BC104132.1 | 1 | 195 | AAI04133.1 | |
BC104133.1 | 1 | 192 | AAI04134.1 | |
BC115705.1 | 1 | 194 | ||
BC115887.1 | 1 | 188 | ||
BC127136.1 | 1 | 195 | ||
BC127137.1 | 1 | 195 |
More Information
Set Membership: |
Intragenic Exonic |