Assay Details
Target Gene Details
Entrez Gene ID: | 18630 |
Gene Name: | plasmacytoma expressed transcript 2 |
Gene Aliases: |
PC231, PC326, Pex3 |
Location: |
Chr.X:89403848-89409689 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pet2 | NM_008821.2 | 12 | 2762 | NP_032847.2 |
BC138080.1 | 11 | 2583 | ||
M95564.1 | 12 | 2767 | AAA39895.1 |
More Information
Set Membership: |
Intragenic Exonic |