Assay Details
Target Gene Details
Entrez Gene ID: | 108673 |
Gene Name: | coiled-coil domain containing 86 |
Gene Aliases: |
4933411H20Rik, 6720480F16Rik, D19Ertd678e |
Location: |
Chr.19:10941481-10949266 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccdc86 | NM_023731.3 | 4 | 2704 | NP_076220.2 |
AK036812.1 | 4 | 2695 |
Target Gene Details
Entrez Gene ID: | 14764 |
Gene Name: | prostaglandin D2 receptor 2 |
Gene Aliases: |
Crth2, Gpr44, Grp45 |
Location: |
Chr.19:10927861-10942511 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptgdr2 | NM_009962.3 | 3 | 1913 | NP_034092.1 |
AB109092.1 | 3 | 1913 | BAC81437.1 | |
AK041947.1 | 3 | 1911 | ||
BC111906.1 | 3 | 1893 | AAI11907.1 |
More Information
Set Membership: |
Intragenic Exonic |