Assay Details
Target Gene Details
Entrez Gene ID: | 22294 |
Gene Name: | ubiquitously expressed transcript |
Gene Aliases: |
0910002B17Rik |
Location: |
Chr.X:20951665-20962035 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Uxt | NM_013840.3 | 7 | 770 | NP_038868.2 |
AK027917.1 | 7 | 771 | BAC25665.1 |
More Information
Set Membership: |
Intragenic Exonic |