Assay Details
Target Gene Details
Entrez Gene ID: | 18169 |
Gene Name: | neuropeptide Y receptor Y6 |
Gene Aliases: |
- |
Location: |
Chr.18:44270127-44277700 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Npy6r | NM_010935.3 | 2 | 1390 | NP_035065.1 |
AK030279.1 | 2 | 1392 | BAC26875.1 | |
BC103620.1 | 2 | 1248 | AAI03621.1 | |
BC103621.1 | 2 | 1248 | AAI03622.1 | |
BC103622.1 | 2 | 1248 | AAI03623.1 | |
BC103667.1 | 2 | 1248 | AAI03668.1 |
More Information
Set Membership: |
Intragenic Exonic |