Assay Details
Target Gene Details
Entrez Gene ID: | 69310 |
Gene Name: | PARK2 co-regulated |
Gene Aliases: |
1700008H23Rik |
Location: |
Chr.17:10403012-10840191 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pacrg | NM_027032.2 | 5 | 1279 | NP_081308.1 |
AK135510.1 | 5 | 1281 | BAE22559.1 |