Assay Details
Target Gene Details
Entrez Gene ID: | 224792 |
Gene Name: | adhesion G protein-coupled receptor F5 |
Gene Aliases: |
8430401C09Rik, 9330185D23, AI528491, Gpr116, mKIAA0758 |
Location: |
Chr.17:43360329-43459557 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adgrf5 | NM_001081178.1 | 17 | 3573 | NP_001074647.1 |
AK173020.1 | 11 | 2692 | BAD32298.1 |