Assay Details
Target Gene Details
Entrez Gene ID: | 13026 |
Gene Name: | phosphate cytidylyltransferase 1, choline, alpha isoform |
Gene Aliases: |
CTalpha, Cctalpha, Ctpct, Cttalpha |
Location: |
Chr.16:32430921-32475065 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pcyt1a | NM_001163159.1 | 1 | 179 | NP_001156631.1 |
NM_001163160.1 | NP_001156632.1 | |||
NM_009981.4 | NP_034111.1 | |||
AK044585.1 | ||||
AK076050.1 | BAC36148.1 | |||
AK076830.1 | BAC36497.1 | |||
BB642590.1 | ||||
L28956.1 | AAA53526.1 | |||
Z12302.1 | CAA78172.1 |
More Information
Set Membership: |
Intragenic Exonic |