Assay Details
Target Gene Details
Entrez Gene ID: | 353344 |
Gene Name: | opsin 5 |
Gene Aliases: |
Gpr136, Neuropsin, PGR12, TMEM13 |
Location: |
Chr.17:42556779-42620470 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Opn5 | NM_181753.4 | 4 | 647 | NP_861418.2 |
AY255592.1 | 1 | 184 | AAO85104.1 | |
AY288426.1 | 4 | 634 | AAP72135.1 | |
AY318865.1 | 4 | 648 | AAR08201.1 | |
BC125611.1 | 4 | 729 | ||
BC132010.1 | 4 | 729 |
More Information
Set Membership: |
Intragenic Exonic |