Assay Details
Target Gene Details
Entrez Gene ID: | 74143 |
Gene Name: | optic atrophy 1 |
Gene Aliases: |
1200011N24Rik, AI225888, AI847218, lilr3, mKIAA0567 |
Location: |
Chr.16:29579306-29663122 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Opa1 | NM_001199177.1 | 2 | 496 | NP_001186106.1 |
NM_133752.3 | 2 | 496 | NP_598513.1 | |
AB044138.1 | 2 | 488 | BAB59000.1 | |
AK004715.1 | 2 | 451 | ||
AK029157.1 | 2 | 454 | BAC26331.1 | |
AK030835.1 | 2 | 450 | ||
AK038446.1 | 2 | 443 | BAC30002.1 | |
AK044657.1 | 2 | 429 | BAC32021.1 | |
AK050383.1 | 2 | 436 | BAC34224.1 | |
AK053861.1 | 2 | 454 | BAC35561.1 | |
AK122311.1 | 2 | 424 | BAC65593.1 | |
AK145620.1 | 2 | 448 | BAE26544.1 | |
BC138665.1 | 2 | 377 | ||
BC145959.1 | 2 | 377 | ||
CB249413.1 | 2 | 497 |
More Information
Set Membership: |
Intragenic Exonic |