Assay Details
Target Gene Details
Entrez Gene ID: | 20583 |
Gene Name: | snail family zinc finger 2 |
Gene Aliases: |
Slug, Slugh, Snail2 |
Location: |
Chr.16:14705859-14709385 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Snai2 | NM_011415.2 | 3 | 1380 | NP_035545.1 |
AK133968.1 | 3 | 1388 | BAE21961.1 | |
BC062164.1 | 3 | 1380 | AAH62164.1 | |
U79550.1 | 3 | 1308 | AAB38365.1 |
More Information
Set Membership: |
Intragenic Exonic |