Assay Details
Target Gene Details
Entrez Gene ID: | 224022 |
Gene Name: | solute carrier family 7 (cationic amino acid transporter, y+ system), member 4 |
Gene Aliases: |
AI853530 |
Location: |
Chr.16:17572018-17576892 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc7a4 | NM_144852.3 | 5 | 2999 | NP_659101.2 |
AK043772.1 | 5 | 3000 | BAC31651.1 |
More Information
Set Membership: |
Intragenic Exonic |