Assay Details
Target Gene Details
Entrez Gene ID: | 224022 |
Gene Name: | solute carrier family 7 (cationic amino acid transporter, y+ system), member 4 |
Gene Aliases: |
AI853530 |
Location: |
Chr.16:17572018-17576892 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc7a4 | NM_144852.3 | 2 | 716 | NP_659101.2 |
AK030586.1 | 2 | 720 | BAC27031.1 | |
AK043772.1 | 2 | 717 | BAC31651.1 | |
AK162398.1 | 2 | 760 | BAE36893.1 | |
BC016100.1 | 2 | 810 | AAH16100.1 |
More Information
Set Membership: |
Intragenic Exonic |