Assay Details
Target Gene Details
Entrez Gene ID: | 110147 |
Gene Name: | euchromatic histone lysine N-methyltransferase 2 |
Gene Aliases: |
Bat8, D17Ertd710e, G9a, KMT1C, NG36 |
Location: |
Chr.17:34898464-34914051 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ehmt2 | NM_001286573.1 | 3 | 566 | NP_001273502.1 |
NM_001286575.1 | 4 | 402 | NP_001273504.1 | |
NM_145830.2 | 3 | 566 | NP_665829.1 | |
NM_147151.2 | 4 | 402 | NP_671493.1 | |
AB077209.1 | 4 | 395 | BAC05482.1 | |
AB077210.1 | 3 | 566 | BAC05483.1 | |
AK154246.1 | 3 | 589 | BAE32462.1 | |
BC058357.1 | 4 | 400 | AAH58357.1 | |
CJ181159.1 |