Assay Details
Target Gene Details
Entrez Gene ID: | 18044 |
Gene Name: | nuclear transcription factor-Y alpha |
Gene Aliases: |
AA407810, Cbf-b, SEZ-10, SEZ10 |
Location: |
Chr.17:48386885-48409901 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nfya | NM_001110832.1 | 10 | 2075 | NP_001104302.1 |
NM_010913.2 | 9 | 1988 | NP_035043.1 | |
AK004729.1 | 10 | 2021 | BAB23511.1 | |
AK143723.1 | 9 | 1950 | BAE25515.1 | |
AK156487.1 | 10 | 1905 | BAE33729.1 | |
BC029695.1 | 9 | 1876 | AAH29695.1 | |
BC057099.1 | 10 | 1977 | AAH57099.1 | |
D78642.1 | 1 | 499 | ||
X55315.1 | 9 | 1898 | CAA39023.1 |
More Information
Set Membership: |
Intragenic Exonic |