Assay Details
Target Gene Details
Entrez Gene ID: | 30940 |
Gene Name: | ubiquitin specific peptidase 25 |
Gene Aliases: |
- |
Location: |
Chr.16:77013789-77116783 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Usp25 | NM_013918.2 | 24 | 3256 | NP_038946.2 |
AF170563.1 | 24 | 3402 | AAF32264.1 | |
AK045966.1 | 6 | 796 | BAC32550.1 | |
AK077769.1 | 9 | 1434 | ||
AK161286.1 | 6 | 826 | BAE36295.1 | |
BC048171.1 | 24 | 3256 | AAH48171.1 | |
BC063059.1 | 24 | 3256 | AAH63059.1 |
More Information
Set Membership: |
Intragenic Exonic |