Assay Details
Target Gene Details
Entrez Gene ID: | 64074 |
Gene Name: | SPARC related modular calcium binding 2 |
Gene Aliases: |
1700056C05Rik, 5430426J21Rik, Smoc2l |
Location: |
Chr.17:14279487-14404790 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smoc2 | NM_022315.2 | 1 | 198 | NP_071710.2 |
AJ249901.1 | 1 | 169 | CAC10355.1 | |
AK030958.1 | 1 | 167 | BAC27193.1 | |
AK040109.1 | 1 | 146 | ||
AK159749.1 | 1 | 165 | BAE35341.1 | |
AK160002.1 | 1 | 165 | BAE35550.1 | |
AK172312.1 | 1 | 163 | ||
BB840063.1 | 1 | 199 | ||
BC019527.1 | 1 | 145 | AAH19527.1 |
More Information
Set Membership: |
Intragenic Exonic |