Assay Details
Target Gene Details
Entrez Gene ID: | 74355 |
Gene Name: | SMC hinge domain containing 1 |
Gene Aliases: |
4931400A14Rik, AW554188, MommeD1, mKIAA0650 |
Location: |
Chr.17:71344489-71475343 on Build GRCm38 |
Assay Gene Location: | Within Exon 48 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smchd1 | NM_028887.3 | 48 | 6329 | NP_083163.3 |
AK016419.1 | 23 | 2806 | BAB30222.1 | |
AK030247.1 | 16 | 1911 | ||
AK129181.1 | 26 | 3326 | BAC97991.1 | |
AK204889.1 | 1 | 138 | ||
BC044905.1 | 12 | 1476 | AAH44905.1 | |
BC058205.1 | 14 | 1789 | AAH58205.1 | |
BC058618.1 | 24 | 2981 | AAH58618.1 | |
BC062946.1 | 34 | 4174 | AAH62946.1 |