Assay Details
Target Gene Details
Entrez Gene ID: | 19876 |
Gene Name: | roundabout homolog 1 (Drosophila) |
Gene Aliases: |
AW494633, AW742721, DUTT1, Gm310 |
Location: |
Chr.16:72027553-73047491 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Robo1 | NM_019413.2 | 2 | 1098 | NP_062286.2 |
AK032752.1 | 2 | 1064 | ||
AK138485.1 | 2 | 1098 | BAE23679.1 | |
AK142047.1 | 2 | 1044 | BAE24924.1 | |
AK143001.1 | 2 | 1098 | BAE25241.1 | |
Y17793.1 | 2 | 360 | CAA76850.1 |
More Information
Set Membership: |
Intragenic Exonic |