Assay Details
Target Gene Details
Entrez Gene ID: | 224727 |
Gene Name: | BCL2-associated athanogene 6 |
Gene Aliases: |
2410045D21Rik, AA408914, BAG-6, Bat3, D17H6S52E, G3, Scythe |
Location: |
Chr.17:35134600-35147322 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bag6 | NM_001252468.1 | 6 | 888 | NP_001239397.1 |
NM_001252469.1 | 6 | 694 | NP_001239398.1 | |
NM_057171.2 | 7 | 942 | NP_476512.1 | |
AK148796.1 | 6 | 808 | BAE28666.1 | |
AK164266.1 | 6 | 665 | ||
AK170134.1 | 7 | 762 | BAE41587.1 | |
BC037051.1 | 7 | 766 | AAH37051.1 | |
BM951977.1 | 3 | 210 |
More Information
Set Membership: |
Intragenic Exonic |