Assay Details
Target Gene Details
Entrez Gene ID: | 239719 |
Gene Name: | MKL/myocardin-like 2 |
Gene Aliases: |
ENSMUSG00000075401, Gt4-1, MRTF-B, Mrtfb, mKIAA1243 |
Location: |
Chr.16:13256017-13417529 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mkl2 | NM_001122667.2 | NP_001116139.1 | ||
NM_153588.3 | 1 | 112 | NP_705816.2 | |
AK158754.1 | 1 | 111 | ||
CJ126058.1 | 1 | 114 |
More Information
Set Membership: |
Intragenic Exonic |