Assay Details
Target Gene Details
Entrez Gene ID: | 20519 |
Gene Name: | solute carrier family 22 (organic cation transporter), member 3 |
Gene Aliases: |
EMT, Oct3, Orct3, Slca22a3 |
Location: |
Chr.17:12419972-12507704 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc22a3 | NM_011395.2 | 11 | 3250 | NP_035525.1 |
AF078750.1 | 11 | 3250 | AAD20978.1 | |
AF082566.1 | 11 | 3133 | AAD53007.1 | |
BC137661.1 | 11 | 3125 | ||
BC137662.1 | 11 | 3125 |
More Information
Set Membership: |
Intragenic Exonic |