Assay Details
Target Gene Details
Entrez Gene ID: | 224860 |
Gene Name: | phospholipase C-like 2 |
Gene Aliases: |
PLC-L2, PRIP-2, Plce2, mKIAA1092 |
Location: |
Chr.17:50508633-50688494 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plcl2 | NM_013880.3 | 6 | 3401 | NP_038908.2 |
AB033615.1 | 6 | 3529 | BAA89457.1 | |
AK134985.1 | 5 | 1732 | ||
AK154288.1 | 6 | 3357 | BAE32490.1 | |
AK165548.1 | 6 | 3407 | ||
BC027746.1 | 6 | 3463 | AAH27746.1 |
More Information
Set Membership: |
Intragenic Exonic |