Assay Details
Target Gene Details
Entrez Gene ID: | 224938 |
Gene Name: | praja 2, RING-H2 motif containing |
Gene Aliases: |
AI447901, AL022700, Neurodap1, mKIAA0438 |
Location: |
Chr.17:64281006-64331883 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pja2 | NM_001025309.1 | 4 | 1109 | NP_001020480.1 |
NM_144859.2 | 4 | 1109 | NP_659108.1 | |
AF493070.1 | 4 | 1166 | AAO85470.1 | |
AK122282.1 | 4 | 1068 | BAC65564.1 | |
AK144586.1 | 4 | 1096 | BAE25949.1 | |
AK164025.1 | 4 | 1114 | ||
AK168371.1 | 4 | 1091 | BAE40303.1 | |
BC017130.1 | 3 | 1035 | AAH17130.1 |
More Information
Set Membership: |
Intragenic Exonic |