Assay Details
Target Gene Details
Entrez Gene ID: | 109801 |
Gene Name: | glyoxalase 1 |
Gene Aliases: |
0610009E22Rik, 1110008E19Rik, 2510049H23Rik, AW550643, GLY1, Glo-1, Glo-1r, Glo-1s, Glo1-r, Glo1-s, Qglo |
Location: |
Chr.17:30592861-30612659 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Glo1 | NM_001113560.1 | NP_001107032.1 | ||
NM_025374.3 | 6 | 1489 | NP_079650.3 | |
AK002386.1 | 6 | 1417 | BAB22060.1 | |
AK003567.1 | 6 | 1437 | BAB22863.1 | |
AK049703.1 | 6 | 1446 | BAC33882.1 | |
BC024663.1 | 6 | 1448 | AAH24663.1 | |
BG077218.2 | ||||
BP769280.1 |
More Information
Set Membership: |
Intragenic Exonic |