Assay Details
Target Gene Details
Entrez Gene ID: | 239985 |
Gene Name: | AT rich interactive domain 1B (SWI-like) |
Gene Aliases: |
8030481M12, 9330189K18Rik, AI836955, Ardi1b, B230217J03Rik, mKIAA1235 |
Location: |
Chr.17:4992755-5347656 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arid1b | NM_001085355.1 | 1 | 1180 | NP_001078824.1 |